Assay Details
Target Gene Details
Entrez Gene ID: | 6929 |
Gene Name: | transcription factor 3 |
Gene Aliases: |
AGM8, E2A, E47, ITF1, TCF-3, VDIR, bHLHb21 |
Location: |
Chr.19:1609290-1652590 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv32942 | Chr.19:1320268 - 1999205 on Build GRCh38 | Gain+Loss | MEX3D APC2 DAZAP1 TCF3 ABHD17A PCSK4 SCAMP4 GAMT CSNK1G2-AS1 UQCR11 REEP6 ONECUT3 ATP8B3 NDUFS7 CSNK1G2 BTBD2 MUM1 C19orf25 MIR1909 ADAT3 LOC100288123 REXO1 ADAMTSL5 PLK5 MBD3 RPS15 KLF16 |
nsv953942 | Chr.19:1371602 - 2057401 on Build GRCh38 | Deletion | MEX3D APC2 DAZAP1 TCF3 ABHD17A MKNK2 PCSK4 SCAMP4 GAMT CSNK1G2-AS1 UQCR11 REEP6 ONECUT3 ATP8B3 NDUFS7 CSNK1G2 BTBD2 MUM1 C19orf25 MIR1909 ADAT3 LOC100288123 REXO1 ADAMTSL5 PLK5 MBD3 RPS15 KLF16 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |