Assay Details
Target Gene Details
Entrez Gene ID: | 10170 |
Gene Name: | dehydrogenase/reductase 9 |
Gene Aliases: |
3-alpha-HSD, 3ALPHA-HSD, RDH-E2, RDH-TBE, RDH15, RDHL, RDHTBE, RETSDR8, SDR9C4 |
Location: |
Chr.2:169067035-169096167 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DHRS9 | NM_001142270.1 | 4 | 751 | NP_001135742.1 |
NM_001142271.1 | 3 | 474 | NP_001135743.1 | |
NM_001289763.1 | 3 | 661 | NP_001276692.1 | |
NM_199204.1 | 4 | 926 | NP_954674.1 | |
XM_011510475.1 | 5 | 1084 | XP_011508777.1 | |
AF067174.1 | 2 | 376 | AAD32458.1 | |
AF240697.1 | 5 | 466 | AAF82747.1 | |
AF240698.1 | 3 | 493 | AAF82748.1 | |
AF295380.1 | 2 | 369 | AAL37037.1 | |
AF343729.1 | 3 | 522 | AAK37528.1 | |
AF529286.1 | 6 | 1872 | AAN04008.1 | |
AF529287.1 | 4 | 826 | AAN04009.1 | |
AF529288.1 | 2 | 434 | AAN04010.1 | |
AK296625.1 | 3 | 619 | ||
AK313220.1 | 4 | 751 | ||
AY017349.1 | 4 | 926 | AAG49422.1 | |
AY359046.1 | 3 | 479 | AAQ89405.1 | |
BC058883.1 | 3 | 479 | AAH58883.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv528457 | Chr.2:169036909 - 169297164 on Build GRCh38 | Gain | DHRS9 LRP2 |
esv2762961 | Chr.2:169037369 - 169297835 on Build GRCh38 | Gain | DHRS9 LRP2 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |