Assay Details
Target Gene Details
Entrez Gene ID: | 53 |
Gene Name: | acid phosphatase 2, lysosomal |
Gene Aliases: |
LAP |
Location: |
Chr.11:47239302-47248847 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ACP2 | NM_001302489.1 | 10 | 1076 | NP_001289418.1 |
NM_001302490.1 | 9 | 1008 | NP_001289419.1 | |
NM_001302491.1 | 8 | 915 | NP_001289420.1 | |
NM_001302492.1 | 9 | 1033 | NP_001289421.1 | |
NM_001610.3 | 10 | 1104 | NP_001601.1 | |
AK298122.1 | 8 | 870 | ||
AK298439.1 | 10 | 1079 | ||
AK300569.1 | 9 | 1032 | ||
AK300911.1 | ||||
AK301860.1 | 9 | 966 | ||
AK303894.1 | 9 | 1000 | ||
AK308512.1 | 8 | 1323 | ||
AK311431.1 | ||||
AY007137.1 | 7 | 636 | ||
BC003160.1 | 10 | 1049 | AAH03160.1 | |
BC093010.1 | 10 | 1062 | AAH93010.1 | |
X12548.1 | 10 | 1058 | CAA31064.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |