Assay Details
Target Gene Details
Entrez Gene ID: | 57144 |
Gene Name: | p21 (RAC1) activated kinase 5 |
Gene Aliases: |
PAK7 |
Location: |
Chr.20:9537390-9839042 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PAK5 | NM_020341.3 | 11 | 3475 | NP_065074.1 |
NM_177990.2 | 10 | 3369 | NP_817127.1 | |
XM_017027960.1 | 12 | 3584 | XP_016883449.1 | |
XM_017027961.1 | 13 | 3639 | XP_016883450.1 | |
XM_017027962.1 | 12 | 3530 | XP_016883451.1 | |
XM_017027963.1 | 10 | 3543 | XP_016883452.1 | |
XM_017027964.1 | 10 | 3925 | XP_016883453.1 | |
XM_017027965.1 | 10 | 3232 | XP_016883454.1 | |
AB033090.1 | 11 | 3477 | ||
AB040812.1 | 11 | 3310 | BAA94194.1 | |
AL133002.1 | 3 | 1187 | CAB61350.1 | |
AX776013.1 | 11 | 3477 | CAE11706.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |