Assay Details
Target Gene Details
Entrez Gene ID: | 23033 |
Gene Name: | dopey family member 1 |
Gene Aliases: |
DOP1, KIAA1117, dJ202D23.2 |
Location: |
Chr.6:83067650-83171351 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DOPEY1 | NM_001199942.1 | 3 | 271 | NP_001186871.1 |
NM_015018.3 | 3 | 271 | NP_055833.2 | |
XM_011535619.2 | 3 | 266 | XP_011533921.1 | |
XM_017010559.1 | 3 | 266 | XP_016866048.1 | |
XM_017010560.1 | 2 | 194 | XP_016866049.1 | |
XM_017010561.1 | 3 | 245 | XP_016866050.1 | |
XM_017010562.1 | 2 | 194 | XP_016866051.1 | |
XM_017010563.1 | 3 | 245 | XP_016866052.1 | |
XM_017010564.1 | 3 | 245 | XP_016866053.1 | |
XM_017010565.1 | 3 | 266 | XP_016866054.1 | |
XM_017010566.1 | 3 | 266 | XP_016866055.1 | |
XM_017010567.1 | 2 | 194 | XP_016866056.1 | |
XM_017010568.1 | 3 | 266 | XP_016866057.1 | |
XM_017010569.1 | 2 | 183 | XP_016866058.1 | |
XM_017010570.1 | 3 | 266 | XP_016866059.1 | |
XM_017010571.1 | 3 | 266 | XP_016866060.1 | |
XM_017010572.1 | 3 | 266 | XP_016866061.1 | |
XM_017010573.1 | 3 | 266 | XP_016866062.1 | |
XM_017010574.1 | 3 | 266 | XP_016866063.1 | |
AK094766.1 | 2 | 173 | ||
BC144544.1 | 2 | 169 | ||
BC150619.1 | 2 | 169 | ||
BX648667.1 | 3 | 271 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1026095 | Chr.6:83040907 - 83209453 on Build GRCh38 | Gain | RWDD2A UBE3D PGM3 DOPEY1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |