Assay Details
Target Gene Details
Entrez Gene ID: | 5653 |
Gene Name: | kallikrein related peptidase 6 |
Gene Aliases: |
Bssp, Klk7, PRSS18, PRSS9, SP59, hK6 |
Location: |
Chr.19:50958631-50969673 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KLK6 | NM_001012964.2 | 6 | 1170 | NP_001012982.1 |
NM_001012965.2 | 5 | 1122 | NP_001012983.1 | |
NM_001319948.1 | 6 | 816 | NP_001306877.1 | |
NM_001319949.1 | 5 | 1013 | NP_001306878.1 | |
NM_002774.3 | 7 | 973 | NP_002765.1 | |
AF013988.1 | 7 | 875 | AAB66483.1 | |
AK314897.1 | ||||
AY279383.1 | 6 | 817 | ||
AY318867.1 | 6 | 979 | AAP82446.1 | |
AY318868.1 | ||||
AY318869.1 | 6 | 965 | AAP82448.1 | |
AY318870.1 | ||||
AY457039.1 | AAR23148.1 | |||
BC015525.2 | 6 | 946 | AAH15525.1 | |
BT006852.1 | AAP35498.1 | |||
D78203.1 | 7 | 884 | BAA11306.1 | |
DQ223012.1 | ABB04464.1 | |||
U62801.1 | 7 | 974 | AAB07113.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs77760094] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
Intragenic Exonic non-DGV Variation |