Assay Details
Target Gene Details
Entrez Gene ID: | 79109 |
Gene Name: | mitogen-activated protein kinase associated protein 1 |
Gene Aliases: |
JC310, MIP1, SIN1, SIN1b, SIN1g |
Location: |
Chr.9:125437394-125707234 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MAPKAP1 | NM_001006617.1 | 12 | 1887 | NP_001006618.1 |
NM_001006619.1 | 11 | 1746 | NP_001006620.1 | |
NM_001006620.1 | 11 | 1559 | NP_001006621.1 | |
NM_001006621.1 | 10 | 1469 | NP_001006622.1 | |
NM_024117.3 | 11 | 1779 | NP_077022.1 | |
XM_011519005.1 | 13 | 2028 | XP_011517307.1 | |
XM_011519006.2 | 12 | 1920 | XP_011517308.1 | |
XM_011519007.1 | 12 | 1887 | XP_011517309.1 | |
XM_011519010.2 | 9 | 1205 | XP_011517312.1 | |
XM_017015126.1 | 12 | 1776 | XP_016870615.1 | |
XM_017015127.1 | 11 | 1668 | XP_016870616.1 | |
AK290060.1 | 11 | 1592 | ||
AK295364.1 | 9 | 1193 | ||
AK298841.1 | 7 | 853 | ||
AK310361.1 | 9 | 1248 | ||
AK316149.1 | 9 | 1132 | ||
AL833042.1 | 11 | 1520 | CAH56311.1 | |
AY524429.1 | 12 | 1884 | AAS90839.1 | |
AY524431.1 | 11 | 1776 | AAS90841.1 | |
AY524432.1 | 11 | 1743 | AAS90842.1 | |
AY633624.1 | 11 | 1746 | AAT46478.1 | |
AY633625.1 | 12 | 1861 | AAT46479.1 | |
AY633626.1 | 13 | 1984 | AAT46480.1 | |
BC002326.2 | 11 | 1748 | AAH02326.1 | |
BC003044.1 | 11 | 1748 | AAH03044.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831722 | Chr.9:125302099 - 125476079 on Build GRCh38 | Gain | GAPVD1 MAPKAP1 |
nsv1046626 | Chr.9:125359313 - 125470470 on Build GRCh38 | Gain | GAPVD1 MAPKAP1 |
nsv515979 | Chr.9:125388505 - 125465747 on Build GRCh38 | Loss | MAPKAP1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |