Assay Details
Target Gene Details
Entrez Gene ID: | 5737 |
Gene Name: | prostaglandin F receptor |
Gene Aliases: |
FP |
Location: |
Chr.1:78490974-78540701 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PTGFR | NM_000959.3 | 2 | 774 | NP_000950.1 |
NM_001039585.1 | 2 | 774 | NP_001034674.1 | |
XM_017001873.1 | 2 | 843 | XP_016857362.1 | |
AF004021.1 | 2 | 659 | AAB63152.1 | |
AK292845.1 | 2 | 758 | ||
AK312816.1 | 2 | 758 | ||
AY337000.1 | 1 | 508 | AAQ76788.1 | |
AY485530.1 | 2 | 745 | AAR84381.1 | |
BC112965.2 | 2 | 685 | AAI12966.1 | |
CD013876.1 | 2 | 778 | ||
L24470.1 | 2 | 745 | AAA17684.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1566 | Chr.1:78486860 - 78495143 on Build GRCh38 | Insertion | PTGFR |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |