Assay Details
Target Gene Details
Entrez Gene ID: | 55268 |
Gene Name: | enoyl-CoA hydratase domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.1:52895910-52927212 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Intron 9 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1001535 | Chr.1:52620368 - 53346539 on Build GRCh38 | Gain | SLC1A7 MAGOH ZYG11B LRP8 MIR5095 MIR1273G COA7 LOC100507564 ZYG11A FAM159A C1orf123 PODN ECHDC2 LOC105378732 MIR1273F CPT2 SCP2 |
nsv829859 | Chr.1:52723470 - 52925273 on Build GRCh38 | Loss | ZYG11A ECHDC2 ZYG11B |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |