Assay Details
Target Gene Details
Entrez Gene ID: | 10499 |
Gene Name: | nuclear receptor coactivator 2 |
Gene Aliases: |
GRIP1, KAT13C, NCoA-2, SRC2, TIF2, bHLHe75 |
Location: |
Chr.8:70109762-70405390 on Build GRCh38 |
Assay Gene Location: | Within Exon 28 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NCOA2 | NM_001321703.1 | 20 | 4298 | NP_001308632.1 |
NM_001321707.1 | 20 | 4144 | NP_001308636.1 | |
NM_001321711.1 | 19 | 4019 | NP_001308640.1 | |
NM_001321712.1 | 19 | 3936 | NP_001308641.1 | |
NM_001321713.1 | 19 | 4039 | NP_001308642.1 | |
NM_006540.3 | 20 | 4226 | NP_006531.1 | |
XM_017012961.1 | 20 | 4420 | XP_016868450.1 | |
XM_017012962.1 | 20 | 4444 | XP_016868451.1 | |
XM_017012963.1 | 20 | 5657 | XP_016868452.1 | |
XM_017012964.1 | 20 | 4712 | XP_016868453.1 | |
XM_017012965.1 | 20 | 4377 | XP_016868454.1 | |
XM_017012966.1 | 21 | 4641 | XP_016868455.1 | |
XM_017012967.1 | 19 | 4355 | XP_016868456.1 | |
XM_017012968.1 | 21 | 4442 | XP_016868457.1 | |
XM_017012969.1 | 19 | 4473 | XP_016868458.1 | |
XM_017012970.1 | 19 | 10892 | XP_016868459.1 | |
XM_017012971.1 | 19 | 4604 | XP_016868460.1 | |
XM_017012972.1 | 16 | 3935 | XP_016868461.1 | |
AK298528.1 | 8 | 1311 | ||
BC114383.1 | 19 | 4093 | ||
X97674.1 | 20 | 4171 | CAA66263.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831352 | Chr.8:69999548 - 70182606 on Build GRCh38 | Gain | NCOA2 PRDM14 |
nsv6247 | Chr.8:70118114 - 70163017 on Build GRCh38 | Deletion | NCOA2 |
nsv6246 | Chr.8:70106516 - 70137048 on Build GRCh38 | Insertion | NCOA2 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |