Assay Details
Target Gene Details
Entrez Gene ID: | 8880 |
Gene Name: | far upstream element binding protein 1 |
Gene Aliases: |
FBP, FUBP, hDH V |
Location: |
Chr.1:77944055-77979494 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FUBP1 | NR_130152.1 | |||
XM_011542391.2 | XP_011540693.1 | |||
XM_011542392.2 | XP_011540694.1 | |||
XM_011542393.2 | XP_011540695.1 | |||
XM_017002739.1 | XP_016858228.1 | |||
XM_017002740.1 | XP_016858229.1 | |||
XM_017002741.1 | XP_016858230.1 | |||
XM_017002742.1 | 21 | 2662 | XP_016858231.1 | |
XM_017002743.1 | XP_016858232.1 | |||
XM_017002744.1 | 20 | 2603 | XP_016858233.1 | |
AB209366.1 | 13 | 2426 | BAD92603.1 | |
BC017247.2 | AAH17247.1 | |||
BX538272.1 | 1 | 129 | ||
CN291186.1 | 1 | 377 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |