Assay Details
Target Gene Details
Entrez Gene ID: | 51657 |
Gene Name: | serine/threonine/tyrosine interacting-like 1 |
Gene Aliases: |
DUSP24, MK-STYX, MKSTYX |
Location: |
Chr.7:75995820-76048709 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
STYXL1 | NM_001317785.1 | NP_001304714.1 | ||
NM_001317786.1 | NP_001304715.1 | |||
NM_001317787.1 | NP_001304716.1 | |||
NM_001317788.1 | NP_001304717.1 | |||
NM_001317789.1 | NP_001304718.1 | |||
NM_016086.2 | NP_057170.1 | |||
NR_134486.1 | ||||
NR_134487.1 | ||||
XM_011516292.2 | 8 | 1131 | XP_011514594.1 | |
XM_011516293.2 | XP_011514595.1 | |||
XM_011516300.1 | XP_011514602.1 | |||
XM_017012296.1 | XP_016867785.1 | |||
XM_017012297.1 | 8 | 1134 | XP_016867786.1 | |
XM_017012298.1 | 8 | 1094 | XP_016867787.1 | |
XM_017012299.1 | XP_016867788.1 | |||
XM_017012300.1 | XP_016867789.1 | |||
XM_017012301.1 | XP_016867790.1 | |||
XM_017012302.1 | XP_016867791.1 | |||
AA782050.1 | ||||
AF069762.1 | AAD36983.1 | |||
AF188202.1 | AAF04112.1 | |||
AF188203.1 | AAF04113.1 | |||
AF188204.1 | AAF04114.1 | |||
AI676056.1 | ||||
AY927581.1 | ||||
BC024035.2 | AAH24035.1 | |||
BM972220.1 | ||||
BX537896.1 | CAD97887.1 | |||
W58723.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv428173 | Chr.7:75934531 - 76173362 on Build GRCh38 | Gain | SNORA14A TMEM120A LOC107986711 GTF2IP7 MDH2 STYXL1 POR |
nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain | HSPB1 LOC441259 STAG3L1 CCL26 LOC105375355 LOC100133091 NSUN5P1 CCL24 LOC541473 MIR4651 STYXL1 RHBDD2 SRRM3 FDPSP2 PMS2P2 SPDYE16 GTF2IRD2B MDH2 POMZP3 GTF2IP7 SPDYE5 TRIM73 GTF2IP1 HIP1 POM121C SSC4D SNORA14A DTX2 TMEM120A LOC107986711 ZP3 PMS2P10 NCF1C PMS2P3 YWHAG POR |
esv3613746 | Chr.7:75965095 - 76003876 on Build GRCh38 | Loss | TMEM120A STYXL1 POR |
esv3613748 | Chr.7:75992925 - 75999800 on Build GRCh38 | Loss | TMEM120A STYXL1 |
esv3613742 | Chr.7:75912784 - 76036530 on Build GRCh38 | Loss | SNORA14A TMEM120A MIR4651 STYXL1 POR |
dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion | HSPB1 CCL26 LOC105375355 LOC100133091 CCL24 MIR4651 STYXL1 RHBDD2 SRRM3 FDPSP2 SPDYE16 MDH2 POMZP3 GTF2IP7 SPDYE5 HIP1 POM121C SSC4D SNORA14A DTX2 TMEM120A LOC107986711 ZP3 PMS2P3 YWHAG POR |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |