Assay Details
Target Gene Details
Entrez Gene ID: | 64121 |
Gene Name: | Ras related GTP binding C |
Gene Aliases: |
GTR2, RAGC, TIB929 |
Location: |
Chr.1:38838197-38859823 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RRAGC | NM_001271851.1 | 7 | 1357 | NP_001258780.1 |
NM_022157.3 | 7 | 1459 | NP_071440.1 | |
AF131773.1 | ||||
AF272035.1 | 7 | 1287 | AAG32662.1 | |
AF323609.1 | 7 | 1282 | AAG45221.1 | |
AK023373.1 | 3 | 487 | BAB14548.1 | |
AK092808.1 | 2 | 2370 | ||
AK298573.1 | 6 | 982 | ||
AK298788.1 | 7 | 1253 | ||
AW327252.1 | 1 | 164 | ||
BC016668.1 | 7 | 1304 | AAH16668.1 | |
CA449149.1 | 1 | 141 | ||
CX761032.1 | 2 | 369 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |