Assay Details
Target Gene Details
Entrez Gene ID: | 55008 |
Gene Name: | HECT and RLD domain containing E3 ubiquitin protein ligase family member 6 |
Gene Aliases: |
- |
Location: |
Chr.4:88378565-88443097 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HERC6 | NM_001165136.1 | NP_001158608.1 | ||
NM_017912.3 | NP_060382.3 | |||
XM_005263083.3 | XP_005263140.1 | |||
XM_011532053.2 | XP_011530355.1 | |||
XM_017008333.1 | XP_016863822.1 | |||
AF336798.1 | AAQ14893.1 | |||
AK092403.1 | BAC03879.1 | |||
AK097168.1 | ||||
AK295832.1 | ||||
AY653201.1 | AAV66895.1 | |||
AY653202.1 | AAV66896.1 | |||
AY653203.1 | AAV66897.1 | |||
BC042047.1 | AAH42047.2 | |||
BX647121.1 | CAI46056.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |