Assay Details
Target Gene Details
Entrez Gene ID: | 10600 |
Gene Name: | ubiquitin specific peptidase 16 |
Gene Aliases: |
UBP-M, UBPM |
Location: |
Chr.21:29024617-29054488 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
USP16 | NM_001001992.1 | 2 | 212 | NP_001001992.1 |
NM_001032410.1 | 3 | 275 | NP_001027582.1 | |
NM_006447.2 | 2 | 212 | NP_006438.1 | |
XM_017028257.1 | 3 | 359 | XP_016883746.1 | |
XM_017028258.1 | 3 | 241 | XP_016883747.1 | |
XM_017028259.1 | 3 | 213 | XP_016883748.1 | |
XM_017028260.1 | XP_016883749.1 | |||
XM_017028261.1 | 3 | 216 | XP_016883750.1 | |
AF126736.1 | 2 | 138 | AAD20949.1 | |
AK025104.1 | 2 | 178 | ||
AK025989.1 | 2 | 178 | ||
AK222681.1 | 2 | 141 | BAD96401.1 | |
AK222884.1 | 2 | 134 | BAD96604.1 | |
AK294284.1 | ||||
AK300771.1 | ||||
AY333928.1 | 2 | 138 | AAR13293.1 | |
BC030777.2 | 2 | 168 | AAH30777.1 | |
CB960929.1 | 2 | 212 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3893402 | Chr.21:28969569 - 29059824 on Build GRCh38 | Gain | USP16 CCT8 RWDD2B LTN1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |