Assay Details
Target Gene Details
Entrez Gene ID: | 84129 |
Gene Name: | acyl-CoA dehydrogenase family member 11 |
Gene Aliases: |
ACAD-11 |
Location: |
Chr.3:132558138-132660131 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ACAD11 | NM_032169.4 | 17 | 2347 | NP_115545.3 |
NR_132426.1 | 16 | 2957 | ||
NR_132427.1 | ||||
NR_132428.1 | 15 | 2130 | ||
AJ608287.1 | 17 | 2298 | CAE55233.1 | |
AK022654.1 | 10 | 941 | BAB14158.1 | |
AK131265.1 | 18 | 2754 | BAD18443.1 | |
AL832873.1 | CAH56354.1 | |||
AL833721.1 | 16 | 2475 | CAH56228.1 | |
AX803783.1 | 17 | 2204 | CAE75545.1 | |
AX803785.1 | 17 | 1967 | CAE75546.1 | |
BC019607.1 | 12 | 1243 | AAH19607.2 | |
BC125204.1 | 17 | 2282 | ||
BC125205.1 | 15 | 2065 |
Target Gene Details
Entrez Gene ID: | 100532724 |
Gene Name: | NPHP3-ACAD11 readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.3:132558138-132722459 on Build GRCh38 |
Assay Gene Location: | Within Exon 42 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NPHP3-ACAD11 | NR_037804.1 | 42 | 6579 | |
AB082531.1 | 42 | 6579 | BAC02709.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |