Assay Details
Target Gene Details
Entrez Gene ID: | 3579 |
Gene Name: | C-X-C motif chemokine receptor 2 |
Gene Aliases: |
CD182, CDw128b, CMKAR2, IL8R2, IL8RA, IL8RB |
Location: |
Chr.2:218125290-218137253 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CXCR2 | NM_001168298.1 | NP_001161770.1 | ||
NM_001557.3 | NP_001548.1 | |||
XM_005246530.3 | XP_005246587.1 | |||
XM_017003990.1 | XP_016859479.1 | |||
XM_017003992.1 | XP_016859481.1 | |||
AK290906.1 | ||||
AK312664.1 | ||||
BC037961.1 | AAH37961.2 | |||
DA670033.1 | ||||
DA674925.1 | ||||
L19593.1 | AAB59437.1 | |||
U11872.1 | AAA64380.1 | |||
U11873.1 | AAA64381.1 | |||
U11874.1 | AAA64382.1 | |||
U11875.1 | AAA64383.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |