Assay Details
Target Gene Details
Entrez Gene ID: | 11022 |
Gene Name: | tudor and KH domain containing |
Gene Aliases: |
TDRD2 |
Location: |
Chr.1:151769354-151790576 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TDRKH | NM_001083963.1 | NP_001077432.1 | ||
NM_001083964.1 | NP_001077433.1 | |||
NM_001083965.1 | NP_001077434.1 | |||
NM_006862.3 | NP_006853.2 | |||
XM_005244856.4 | XP_005244913.1 | |||
XM_017000122.1 | XP_016855611.1 | |||
XM_017000123.1 | XP_016855612.1 | |||
XM_017000124.1 | XP_016855613.1 | |||
XM_017000125.1 | XP_016855614.1 | |||
XM_017000126.1 | XP_016855615.1 | |||
XM_017000127.1 | XP_016855616.1 | |||
XM_017000128.1 | XP_016855617.1 | |||
XM_017000129.1 | XP_016855618.1 | |||
XM_017000130.1 | XP_016855619.1 | |||
AF119121.2 | AAD30971.1 | |||
AF227192.1 | AAF36701.1 | |||
AK056402.1 | ||||
AK225160.1 | ||||
AK226092.1 | ||||
AK295948.1 | ||||
AK303063.1 | ||||
AK316279.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |