Assay Details
Target Gene Details
Entrez Gene ID: | 64850 |
Gene Name: | ethanolamine-phosphate phospho-lyase |
Gene Aliases: |
AGXT2L1 |
Location: |
Chr.4:108742040-108763180 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ETNPPL | NM_001146590.1 | NP_001140062.1 | ||
NM_001146627.1 | NP_001140099.1 | |||
NM_031279.3 | NP_112569.2 | |||
NR_027474.1 | ||||
NR_027475.1 | ||||
XM_006714290.2 | XP_006714353.1 | |||
AJ298293.1 | CAC22253.1 | |||
AK091888.1 | BAC03766.1 | |||
AK294072.1 | ||||
AK294235.1 | ||||
BC022526.1 | AAH22526.1 | |||
DC311077.1 | ||||
DC327590.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv519226 | Chr.4:108739559 - 108766158 on Build GRCh38 | Loss | ETNPPL |
nsv1000660 | Chr.4:108730447 - 108787621 on Build GRCh38 | Gain | ETNPPL |
nsv830034 | Chr.4:108736535 - 108898569 on Build GRCh38 | Loss | ETNPPL COL25A1 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |