Assay Details
Target Gene Details
Entrez Gene ID: | 7701 |
Gene Name: | zinc finger protein 142 |
Gene Aliases: |
pHZ-49 |
Location: |
Chr.2:218637592-218659632 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZNF142 | NM_001105537.2 | 8 | 2635 | NP_001099007.1 |
NR_073599.1 | 9 | 2944 | ||
NR_073600.1 | 9 | 2777 | ||
XM_006712730.3 | 9 | 3310 | XP_006712793.1 | |
XM_011511784.2 | 9 | 3405 | XP_011510086.1 | |
XM_011511785.2 | 9 | 3330 | XP_011510087.1 | |
XM_011511786.2 | 9 | 3235 | XP_011510088.1 | |
XM_011511787.2 | 8 | 3230 | XP_011510089.1 | |
XM_011511788.2 | 8 | 3135 | XP_011510090.1 | |
XM_011511789.2 | 8 | 2803 | XP_011510091.1 | |
XM_017004871.1 | 8 | 3386 | XP_016860360.1 | |
XM_017004872.1 | 7 | 2786 | XP_016860361.1 | |
AK095027.1 | ||||
D87073.1 | 8 | 2633 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |