Assay Details
Target Gene Details
Entrez Gene ID: | 64852 |
Gene Name: | terminal uridylyl transferase 1, U6 snRNA-specific |
Gene Aliases: |
PAPD2, RBM21, STARPAP, TUTase, URLC6 |
Location: |
Chr.11:62575045-62591637 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TUT1 | NM_022830.2 | NP_073741.2 | ||
AB101208.1 | ||||
AK026000.1 | BAB15314.1 | |||
AK292610.1 | ||||
BC110910.1 | AAI10911.1 | |||
BC128263.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3584599 | Chr.11:62495270 - 62630578 on Build GRCh38 | Gain | AHNAK MIR6747 EML3 TUT1 MTA2 B3GAT3 EEF1G GANAB ROM1 |
nsv832180 | Chr.11:62434133 - 62597356 on Build GRCh38 | Loss | AHNAK MIR6747 TUT1 MTA2 EEF1G |
nsv825939 | Chr.11:62558611 - 62643105 on Build GRCh38 | Gain | MIR6747 EML3 TUT1 MTA2 B3GAT3 EEF1G GANAB ROM1 |
nsv832182 | Chr.11:62528103 - 62695376 on Build GRCh38 | Gain+Loss | UQCC3 AHNAK EML3 SNORA57 METTL12 INTS5 GANAB HNRNPUL2-BSCL2 MIR6747 TUT1 MTA2 BSCL2 B3GAT3 EEF1G LBHD1 UBXN1 C11orf98 LRRN4CL ROM1 |
nsv555174 | Chr.11:62566624 - 62729047 on Build GRCh38 | Gain | UQCC3 EML3 SNORA57 METTL12 GNG3 INTS5 GANAB HNRNPUL2-BSCL2 MIR6747 TTC9C TUT1 MTA2 BSCL2 B3GAT3 EEF1G HNRNPUL2 LBHD1 UBXN1 C11orf98 LRRN4CL ROM1 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |