Assay Details
Target Gene Details
Entrez Gene ID: | 2017 |
Gene Name: | cortactin |
Gene Aliases: |
EMS1 |
Location: |
Chr.11:70398506-70436584 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CTTN | NM_001184740.1 | NP_001171669.1 | ||
NM_005231.3 | NP_005222.2 | |||
NM_138565.2 | NP_612632.1 | |||
XM_006718447.2 | XP_006718510.1 | |||
XM_006718448.3 | XP_006718511.1 | |||
XM_017017312.1 | XP_016872801.1 | |||
AK222613.1 | BAD96333.1 | |||
AK291097.1 | ||||
AU124856.1 | ||||
BC008799.2 | AAH08799.1 | |||
BC033889.1 | AAH33889.1 | |||
DA716772.1 | ||||
M98343.1 | AAA58455.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |