Assay Details
Target Gene Details
Entrez Gene ID: | 51267 |
Gene Name: | C-type lectin domain family 1 member A |
Gene Aliases: |
CLEC-1, CLEC1 |
Location: |
Chr.12:10069504-10099065 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CLEC1A | NM_001297748.1 | 5 | 1485 | NP_001284677.1 |
NM_001297749.1 | 4 | 1308 | NP_001284678.1 | |
NM_001297750.1 | 7 | 1682 | NP_001284679.1 | |
NM_001297751.1 | 6 | 1626 | NP_001284680.1 | |
NM_016511.3 | 6 | 1584 | NP_057595.2 | |
XM_011520687.2 | 7 | 1665 | XP_011518989.1 | |
AF200949.1 | 6 | 1522 | AAF36830.1 | |
AK075114.1 | 6 | 1504 | BAC11410.1 | |
AK091989.1 | 5 | 1436 | ||
AK300979.1 | ||||
AY358587.1 | 6 | 1584 | AAQ88950.1 | |
BC039072.1 | 6 | 1490 | AAH39072.1 | |
BC067746.1 | 6 | 1504 | AAH67746.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |