Assay Details
Target Gene Details
Entrez Gene ID: | 54496 |
Gene Name: | protein arginine methyltransferase 7 |
Gene Aliases: |
- |
Location: |
Chr.16:68310974-68358832 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PRMT7 | XM_011523112.2 | 19 | 2809 | XP_011521414.1 |
XM_011523113.2 | 20 | 2936 | XP_011521415.1 | |
XM_011523115.2 | 19 | 2753 | XP_011521417.1 | |
XM_011523116.2 | 19 | 2806 | XP_011521418.1 | |
XM_011523121.2 | 17 | 2574 | XP_011521423.1 | |
XM_011523124.2 | 19 | 2862 | XP_011521426.1 | |
XM_011523125.2 | 18 | 2704 | XP_011521427.1 | |
XM_011523126.2 | 18 | 2708 | XP_011521428.1 | |
XM_011523128.2 | 19 | 2753 | XP_011521430.1 | |
XM_017023290.1 | 20 | 2933 | XP_016878779.1 | |
XM_017023291.1 | 20 | 2873 | XP_016878780.1 | |
XM_017023292.1 | 19 | 2750 | XP_016878781.1 | |
XM_017023296.1 | 20 | 2959 | XP_016878785.1 | |
XM_017023297.1 | 20 | 2956 | XP_016878786.1 | |
XM_017023298.1 | 18 | 2804 | XP_016878787.1 | |
XM_017023300.1 | 17 | 2714 | XP_016878789.1 | |
XM_017023301.1 | 17 | 2711 | XP_016878790.1 | |
XM_017023302.1 | 18 | 2701 | XP_016878791.1 | |
XM_017023303.1 | 19 | 2859 | XP_016878792.1 | |
XM_017023311.1 | 18 | 2657 | XP_016878800.1 | |
AB067520.1 | 10 | 3312 | BAB67826.1 |
Target Gene Details
Entrez Gene ID: | 55512 |
Gene Name: | sphingomyelin phosphodiesterase 3 |
Gene Aliases: |
NSMASE2 |
Location: |
Chr.16:68358327-68448703 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SMPD3 | NM_018667.3 | NP_061137.1 | ||
XM_005256031.3 | XP_005256088.1 | |||
XM_005256032.3 | XP_005256089.1 | |||
XM_011523207.1 | XP_011521509.1 | |||
XM_011523208.2 | XP_011521510.1 | |||
XM_011523209.2 | XP_011521511.1 | |||
XM_017023405.1 | XP_016878894.1 | |||
XM_017023406.1 | XP_016878895.1 | |||
XM_017023407.1 | XP_016878896.1 | |||
XM_017023408.1 | XP_016878897.1 | |||
AK026246.1 | BAB15409.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3582280 | Chr.16:68348088 - 68360930 on Build GRCh38 | Loss | SMPD3 PRMT7 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic DGV Variation |