Assay Details
Target Gene Details
Entrez Gene ID: | 84364 |
Gene Name: | ADP ribosylation factor GTPase activating protein 2 |
Gene Aliases: |
IRZ, NBLA10535, ZFP289, ZNF289 |
Location: |
Chr.11:47164298-47177503 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ARFGAP2 | NM_001242832.1 | 15 | 2538 | NP_001229761.1 |
NM_032389.4 | 16 | 2622 | NP_115765.2 | |
XM_005253166.1 | 17 | 3117 | XP_005253223.1 | |
XM_005253167.1 | 17 | 3084 | XP_005253224.1 | |
XM_005253168.2 | 17 | 3042 | XP_005253225.1 | |
XM_006718346.1 | 16 | 3075 | XP_006718409.1 | |
XM_011520405.1 | 16 | 2958 | XP_011518707.1 | |
XM_017018413.1 | 16 | 3042 | XP_016873902.1 | |
XM_017018414.1 | 16 | 2438 | XP_016873903.1 | |
XM_017018415.1 | 16 | 2390 | XP_016873904.1 | |
AB073358.1 | 15 | 2232 | BAE45716.1 | |
AK027482.1 | 16 | 2358 | BAB55144.1 | |
AK098324.1 | 11 | 1968 | ||
AK129511.1 | 6 | 1315 | ||
AL834337.1 | 16 | 2341 | CAD39004.1 | |
BC030148.2 | 16 | 2365 | AAH30148.1 | |
CD366084.1 | 1 | 418 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |