Assay Details
Target Gene Details
Entrez Gene ID: | 26289 |
Gene Name: | adenylate kinase 5 |
Gene Aliases: |
AK6 |
Location: |
Chr.1:77281908-77559969 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AK5 | NM_012093.3 | NP_036225.2 | ||
NM_174858.2 | NP_777283.1 | |||
XM_005270739.4 | XP_005270796.1 | |||
XM_006710572.3 | XP_006710635.1 | |||
XM_017001008.1 | XP_016856497.1 | |||
XM_017001009.1 | XP_016856498.1 | |||
XM_017001010.1 | XP_016856499.1 | |||
XM_017001011.1 | XP_016856500.1 | |||
XM_017001012.1 | XP_016856501.1 | |||
AF445193.1 | AAP97322.1 | |||
AK090967.1 | BAC03558.1 | |||
AY171600.2 | AAO16520.2 | |||
BC033896.1 | AAH33896.1 | |||
BC036666.1 | AAH36666.1 | |||
DN993295.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |