Assay Details
Target Gene Details
Entrez Gene ID: | 56834 |
Gene Name: | G protein-coupled receptor 137 |
Gene Aliases: |
C11orf4, GPR137A, TM7SF1L1 |
Location: |
Chr.11:64270337-64289500 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPR137 | NM_001170726.1 | 6 | 893 | NP_001164197.1 |
NM_001170880.1 | 4 | 1106 | NP_001164351.1 | |
NM_001170881.1 | NP_001164352.1 | |||
NM_001177358.1 | 5 | 1158 | NP_001170829.1 | |
NM_020155.3 | 4 | 1106 | NP_064540.3 | |
XM_005274100.2 | 5 | 1121 | XP_005274157.1 | |
XM_005274101.2 | 5 | 1321 | XP_005274158.1 | |
XM_005274102.2 | 5 | 837 | XP_005274159.1 | |
XM_005274104.2 | 5 | 785 | XP_005274161.1 | |
XM_011545168.2 | 5 | 1031 | XP_011543470.1 | |
XM_011545169.1 | 5 | 750 | XP_011543471.1 | |
XM_011545170.2 | 5 | 1031 | XP_011543472.1 | |
XM_011545171.2 | XP_011543473.1 | |||
XM_011545172.2 | 5 | 1031 | XP_011543474.1 | |
XM_017018014.1 | 5 | 1321 | XP_016873503.1 | |
XM_017018015.1 | 5 | 750 | XP_016873504.1 | |
XM_017018016.1 | XP_016873505.1 | |||
AK056094.1 | 5 | 999 | BAB71093.1 | |
AK300209.1 | 6 | 893 | ||
AK302256.1 | 5 | 1158 | ||
BC033920.1 | 4 | 798 | AAH33920.1 | |
BC035002.1 | AAH35002.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv951010 | Chr.11:63942829 - 64352128 on Build GRCh38 | Deletion | LOC105369340 PLCB3 GPR137 TEX40 OTUB1 PRDX5 FLRT1 TRMT112 KCNK4-TEX40 TRPT1 NAA40 NUDT22 ESRRA STIP1 PPP1R14B VEGFB MACROD1 KCNK4 MIR7155 BAD FERMT3 FKBP2 COX8A DNAJC4 CCDC88B |
dgv1963n54 | Chr.11:64174262 - 64329761 on Build GRCh38 | Loss | PPP1R14B VEGFB LOC105369340 PLCB3 GPR137 KCNK4 TEX40 PRDX5 BAD FERMT3 FKBP2 TRMT112 KCNK4-TEX40 TRPT1 DNAJC4 NUDT22 ESRRA STIP1 |
nsv832186 | Chr.11:64204788 - 64402587 on Build GRCh38 | Loss | PPP1R14B VEGFB LOC105369340 PLCB3 GPR137 MIR1237 KCNK4 MIR7155 TEX40 RPS6KA4 PRDX5 BAD FERMT3 FKBP2 TRMT112 KCNK4-TEX40 TRPT1 DNAJC4 NUDT22 ESRRA CCDC88B |
nsv469961 | Chr.11:64055468 - 64318495 on Build GRCh38 | Loss | PPP1R14B VEGFB MACROD1 LOC105369340 PLCB3 GPR137 KCNK4 TEX40 PRDX5 BAD FERMT3 FLRT1 FKBP2 TRMT112 KCNK4-TEX40 TRPT1 DNAJC4 NUDT22 ESRRA STIP1 |
nsv357 | Chr.11:64284335 - 64308789 on Build GRCh38 | Insertion | KCNK4-TEX40 ESRRA GPR137 KCNK4 TEX40 BAD |