Assay Details
Target Gene Details
Entrez Gene ID: | 83416 |
Gene Name: | Fc receptor like 5 |
Gene Aliases: |
BXMAS1, CD307, CD307e, FCRH5, IRTA2, PRO820 |
Location: |
Chr.1:157513377-157552520 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FCRL5 | NM_001195388.1 | 17 | 4813 | NP_001182317.1 |
NM_031281.2 | 17 | 4823 | NP_112571.2 | |
AF343664.1 | 17 | 4758 | AAK31327.1 | |
AF369794.2 | 18 | 4823 | AAK50059.2 | |
AK090423.1 | 9 | 3759 | BAC03404.1 | |
AK090475.1 | 7 | 3942 | BAC03456.1 | |
AL832301.1 | 2 | 2053 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831681 | Chr.1:157425048 - 157527722 on Build GRCh38 | Gain+Loss | FCRL5 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |