Assay Details
Target Gene Details
Entrez Gene ID: | 81887 |
Gene Name: | LAS1 like, ribosome biogenesis factor |
Gene Aliases: |
Las1-like, dJ475B7.2 |
Location: |
Chr.X:65512582-65534806 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LAS1L | NM_001170649.1 | 5 | 840 | NP_001164120.1 |
NM_001170650.1 | 4 | 714 | NP_001164121.1 | |
NM_031206.4 | 5 | 840 | NP_112483.1 | |
XM_005262301.1 | 5 | 825 | XP_005262358.1 | |
XM_005262304.1 | 5 | 825 | XP_005262361.1 | |
XM_005262305.3 | 5 | 825 | XP_005262362.1 | |
XM_005262306.3 | 5 | 825 | XP_005262363.1 | |
XM_011531045.1 | 4 | 689 | XP_011529347.1 | |
XM_011531046.1 | 5 | 825 | XP_011529348.1 | |
XM_017029877.1 | 5 | 825 | XP_016885366.1 | |
XM_017029878.1 | 5 | 825 | XP_016885367.1 | |
XM_017029879.1 | 5 | 825 | XP_016885368.1 | |
XM_017029880.1 | 5 | 825 | XP_016885369.1 | |
AF116730.1 | 5 | 778 | AAO15306.1 | |
AK022587.1 | 5 | 790 | BAB14114.1 | |
AK074087.1 | 5 | 809 | BAB84913.1 | |
AK097383.1 | 4 | 664 | ||
AK225059.1 | 5 | 773 | ||
AK295591.1 | ||||
BC014545.1 | 5 | 824 | AAH14545.1 | |
BC018610.1 | 5 | 788 | AAH18610.1 | |
BC019302.1 | 5 | 785 | AAH19302.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |