Assay Details
Target Gene Details
Entrez Gene ID: | 83892 |
Gene Name: | potassium channel tetramerization domain containing 10 |
Gene Aliases: |
BTBD28, MSTP028, ULRO61, hBACURD3 |
Location: |
Chr.12:109448655-109477544 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KCTD10 | NM_001317395.1 | 7 | 1332 | NP_001304324.1 |
NM_001317399.1 | 7 | 1260 | NP_001304328.1 | |
NM_031954.4 | 7 | 1329 | NP_114160.1 | |
NR_133897.1 | 6 | 1159 | ||
NR_133898.1 | 6 | 1115 | ||
AB209358.1 | 2 | 3874 | BAD92595.1 | |
AF113208.1 | 7 | 1074 | AAG39279.1 | |
AK027543.1 | 7 | 1135 | BAB55188.1 | |
AK056912.1 | 7 | 2737 | ||
AK092170.1 | 6 | 948 | ||
AK123733.1 | 4 | 1566 | ||
AK309783.1 | 6 | 944 | ||
AY205299.1 | 7 | 1135 | AAO47716.1 | |
BC040062.1 | 7 | 1082 | AAH40062.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |