Assay Details
Target Gene Details
Entrez Gene ID: | 9743 |
Gene Name: | Rho GTPase activating protein 32 |
Gene Aliases: |
GC-GAP, GRIT, PX-RICS, RICS, p200RhoGAP, p250GAP |
Location: |
Chr.11:128965060-129279168 on Build GRCh38 |
Assay Gene Location: | Within Exon 28 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ARHGAP32 | NM_001142685.1 | 21 | 3779 | NP_001136157.1 |
NM_014715.3 | 12 | 3111 | NP_055530.2 | |
XM_005271736.4 | 22 | 4809 | XP_005271793.2 | |
XM_011543072.2 | 22 | 4011 | XP_011541374.1 | |
XM_011543073.2 | 22 | 3747 | XP_011541375.2 | |
XM_011543075.2 | 13 | 2957 | XP_011541377.1 | |
XM_017018595.1 | 22 | 3728 | XP_016874084.1 | |
XM_017018596.1 | 20 | 4005 | XP_016874085.1 | |
AB018255.2 | 12 | 3032 | ||
AB088416.1 | 12 | 3032 | ||
AK025237.1 | 1 | 277 | ||
AL833062.2 | 19 | 4272 | CAD89974.1 | |
AY194287.1 | 12 | 3035 | AAO43677.1 | |
BC000277.1 | AAH00277.2 | |||
BC104898.1 | 11 | 2766 | AAI04899.1 | |
BC113429.1 | 11 | 2766 | ||
EF127492.1 | 21 | 3779 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv525837 | Chr.11:128969261 - 128972978 on Build GRCh38 | Loss | ARHGAP32 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |