Assay Details
Target Gene Details
Entrez Gene ID: | 114926 |
Gene Name: | small integral membrane protein 19 |
Gene Aliases: |
C8orf40 |
Location: |
Chr.8:42541155-42552997 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SMIM19 | NM_001135674.1 | 2 | 810 | NP_001129146.1 |
NM_001135675.1 | 2 | 418 | NP_001129147.1 | |
NM_001135676.1 | 2 | 103 | NP_001129148.1 | |
NM_138436.3 | 2 | 189 | NP_612445.2 | |
XM_005273398.4 | 2 | 724 | XP_005273455.1 | |
AK311932.1 | 2 | 164 | ||
BC013035.2 | 2 | 151 | AAH13035.1 | |
BI335341.1 | ||||
BI463786.1 | 2 | 108 | ||
BI549696.1 | 2 | 423 | ||
DA702519.1 | 2 | 189 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1017899 | Chr.8:42510362 - 42586679 on Build GRCh38 | Gain | SMIM19 SLC20A2 |