Assay Details
Target Gene Details
Entrez Gene ID: | 60401 |
Gene Name: | ectodysplasin A2 receptor |
Gene Aliases: |
EDA-A2R, EDAA2R, TNFRSF27, XEDAR |
Location: |
Chr.X:66594384-66639303 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EDA2R | NM_001199687.2 | 7 | 2823 | NP_001186616.1 |
NM_001242310.1 | 7 | 2788 | NP_001229239.1 | |
NM_001324199.1 | 5 | 2469 | NP_001311128.1 | |
NM_001324201.1 | 6 | 2743 | NP_001311130.1 | |
NM_001324202.1 | 5 | 2172 | NP_001311131.1 | |
NM_001324204.1 | 5 | 2271 | NP_001311133.1 | |
NM_001324205.1 | 5 | 2265 | NP_001311134.1 | |
NM_001324206.1 | 7 | 2831 | NP_001311135.1 | |
NM_021783.4 | 7 | 2825 | NP_068555.1 | |
NR_136726.1 | 7 | 2732 | ||
NR_136727.1 | 7 | 2734 | ||
XM_011531000.1 | 8 | 2838 | XP_011529302.1 | |
XM_011531001.1 | 8 | 2886 | XP_011529303.1 | |
XM_011531002.1 | 8 | 2543 | XP_011529304.1 | |
XM_011531003.2 | 6 | 2638 | XP_011529305.1 | |
XM_011531005.2 | 5 | 2472 | XP_011529307.1 | |
XM_011531006.2 | 6 | 2337 | XP_011529308.1 | |
XM_017029703.1 | 6 | 2735 | XP_016885192.1 | |
XM_017029704.1 | 7 | 2480 | XP_016885193.1 | |
XM_017029705.1 | 6 | 2613 | XP_016885194.1 | |
XM_017029706.1 | 6 | 2388 | XP_016885195.1 | |
XM_017029707.1 | 6 | 2522 | XP_016885196.1 | |
XM_017029708.1 | 5 | 2164 | XP_016885197.1 | |
BC034919.1 | 7 | 2772 | AAH34919.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv2305e212 | Chr.X:66550620 - 66717352 on Build GRCh38 | Gain | EDA2R |
nsv520630 | Chr.X:66486237 - 66797185 on Build GRCh38 | Gain+Loss | EDA2R |
esv3576919 | Chr.X:65837678 - 66804698 on Build GRCh38 | Gain | VSIG4 MIR223 HEPH EDA2R |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |