Assay Details
Target Gene Details
Entrez Gene ID: | 23351 |
Gene Name: | KH and NYN domain containing |
Gene Aliases: |
KIAA0323 |
Location: |
Chr.14:24423186-24441342 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KHNYN | NM_001290256.1 | 3 | 1450 | NP_001277185.1 |
NM_001290257.1 | 3 | 1385 | NP_001277186.1 | |
NM_015299.2 | 3 | 1513 | NP_056114.1 | |
XM_005267474.4 | 3 | 1469 | XP_005267531.1 | |
XM_011536590.2 | 3 | 1619 | XP_011534892.1 | |
XM_017021114.1 | 3 | 1385 | XP_016876603.1 | |
AB002321.1 | 3 | 1450 | ||
AK291680.1 | 3 | 1366 | ||
BC015780.1 | 1 | 536 | AAH15780.1 | |
BC042554.1 | 3 | 1478 | AAH42554.1 | |
BX161465.1 | 3 | 1486 | CAD61924.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |