Assay Details
Target Gene Details
Entrez Gene ID: | 4781 |
Gene Name: | nuclear factor I B |
Gene Aliases: |
CTF, HMGIC/NFIB, NF-I/B, NF1-B, NFI-B, NFI-RED, NFIB2, NFIB3 |
Location: |
Chr.9:14081843-14398983 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NFIB | NM_001190737.1 | NP_001177666.1 | ||
NM_001190738.1 | NP_001177667.1 | |||
NM_001282787.1 | NP_001269716.1 | |||
NM_005596.3 | NP_005587.2 | |||
XM_005251467.2 | XP_005251524.1 | |||
XM_005251468.4 | XP_005251525.1 | |||
XM_005251469.2 | XP_005251526.1 | |||
XM_005251470.2 | XP_005251527.1 | |||
XM_005251471.3 | XP_005251528.1 | |||
XM_006716773.2 | XP_006716836.1 | |||
XM_006716774.2 | XP_006716837.1 | |||
XM_006716775.2 | XP_006716838.1 | |||
XM_017014739.1 | XP_016870228.1 | |||
XM_017014740.1 | XP_016870229.1 | |||
XM_017014741.1 | XP_016870230.1 | |||
XM_017014742.1 | XP_016870231.1 | |||
AI140312.1 | ||||
AK094066.1 | ||||
AK129782.1 | ||||
AL110126.1 | ||||
BX537698.1 | ||||
BX537707.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1033764 | Chr.9:13764919 - 14226169 on Build GRCh38 | Loss | NFIB LINC00583 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |