Assay Details
Target Gene Details
Entrez Gene ID: | 80146 |
Gene Name: | UDP-glucuronate decarboxylase 1 |
Gene Aliases: |
SDR6E1, UGD |
Location: |
Chr.2:106093303-106194339 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UXS1 | NM_001253875.1 | 15 | 1616 | NP_001240804.1 |
NM_001253876.1 | 10 | 1753 | NP_001240805.1 | |
NM_025076.4 | 15 | 1601 | NP_079352.2 | |
NR_045607.1 | 14 | 1552 | ||
XM_006712775.3 | 14 | 1474 | XP_006712838.1 | |
XM_011511902.2 | 15 | 1595 | XP_011510204.2 | |
XM_011511903.2 | 15 | 1578 | XP_011510205.2 | |
XM_011511904.2 | 12 | 1923 | XP_011510206.1 | |
XM_017005008.1 | 13 | 1416 | XP_016860497.1 | |
XM_017005009.1 | 13 | 1401 | XP_016860498.1 | |
XM_017005010.1 | 14 | 1459 | XP_016860499.1 | |
XM_017005011.1 | 15 | 1703 | XP_016860500.1 | |
XM_017005012.1 | 15 | 1638 | XP_016860501.1 | |
XM_017005013.1 | 13 | 2161 | XP_016860502.1 | |
XM_017005014.1 | 14 | 1379 | XP_016860503.1 | |
AI217133.1 | ||||
AK027244.1 | 10 | 1753 | BAB15705.1 | |
AK056022.1 | 4 | 2430 | ||
AK075120.1 | 15 | 1563 | BAC11415.1 | |
AK075170.1 | 15 | 1578 | BAC11448.1 | |
AK122696.1 | 14 | 1517 | ||
AK290668.1 | 15 | 1576 | ||
AL834439.1 | 2 | 1365 | CAD39099.2 | |
AY147934.1 | 15 | 1568 | AAN39844.1 | |
AY358541.1 | 15 | 1552 | AAQ88905.1 | |
BC009819.2 | 15 | 1542 | AAH09819.2 | |
BX366726.2 | 1 | 460 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |