Assay Details
Target Gene Details
Entrez Gene ID: | 10911 |
Gene Name: | urotensin 2 |
Gene Aliases: |
PRO1068, U-II, UCN2, UII |
Location: |
Chr.1:7846345-7913615 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UTS2 | NM_006786.3 | 4 | 400 | NP_006777.1 |
NM_021995.2 | 5 | 464 | NP_068835.1 | |
XM_011540537.2 | 6 | 913 | XP_011538839.1 | |
XM_011540538.1 | 5 | 558 | XP_011538840.1 | |
XM_017000116.1 | XP_016855605.1 | |||
XM_017000117.1 | XP_016855606.1 | |||
XM_017000118.1 | XP_016855607.1 | |||
XM_017000119.1 | XP_016855608.1 | |||
AF104118.1 | 4 | 391 | AAD13070.1 | |
AF140630.1 | AAD55577.1 | |||
AY358375.1 | 4 | 400 | AAQ88741.1 | |
BC126443.1 | 4 | 441 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |