Assay Details
Target Gene Details
Entrez Gene ID: | 10713 |
Gene Name: | ubiquitin specific peptidase 39 |
Gene Aliases: |
65K, CGI-21, HSPC332, SAD1, SNRNP65 |
Location: |
Chr.2:85602842-85649284 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
USP39 | NM_001256725.1 | 1 | 336 | NP_001243654.1 |
NM_001256726.1 | 1 | 336 | NP_001243655.1 | |
NM_001256727.1 | NP_001243656.1 | |||
NM_001256728.1 | NP_001243657.1 | |||
NM_006590.3 | 1 | 336 | NP_006581.2 | |
NR_046347.1 | ||||
XM_006711922.1 | XP_006711985.1 | |||
XM_006711923.2 | 1 | 290 | XP_006711986.1 | |
XM_011532487.1 | 1 | 276 | XP_011530789.1 | |
XM_011532488.1 | XP_011530790.1 | |||
XM_017003182.1 | 1 | 273 | XP_016858671.1 | |
AF132955.1 | 1 | 243 | AAD27730.1 | |
AF353989.1 | 1 | 243 | AAK49524.1 | |
AK001047.1 | 1 | 234 | ||
AK056334.1 | ||||
AK129872.1 | ||||
AK222852.1 | 1 | 239 | BAD96572.1 | |
AK289451.1 | 1 | 290 | ||
AK295257.1 | ||||
AK302236.1 | ||||
AK309485.1 | 1 | 139 | ||
AL519827.3 | 1 | 285 | ||
BC001384.2 | 1 | 225 | AAH01384.2 | |
BC067273.1 | 1 | 264 | AAH67273.1 | |
BX462623.2 | 1 | 268 | ||
DA548921.1 | ||||
DB124294.1 | 1 | 336 | ||
DB444135.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |