Assay Details
Target Gene Details
Entrez Gene ID: | 115350 |
Gene Name: | Fc receptor like 1 |
Gene Aliases: |
CD307a, FCRH1, IFGP1, IRTA5 |
Location: |
Chr.1:157792249-157820150 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FCRL1 | NM_001159397.1 | 3 | 265 | NP_001152869.1 |
NM_001159398.1 | 3 | 265 | NP_001152870.1 | |
NM_052938.4 | 3 | 265 | NP_443170.1 | |
XM_005244866.2 | 3 | 236 | XP_005244923.1 | |
XM_005244867.2 | 3 | 236 | XP_005244924.1 | |
XM_005244869.3 | XP_005244926.1 | |||
XM_011509125.1 | 3 | 236 | XP_011507427.1 | |
XM_011509126.1 | 3 | 236 | XP_011507428.1 | |
XM_011509127.2 | 2 | 228 | XP_011507429.1 | |
XM_011509128.1 | 3 | 236 | XP_011507430.1 | |
XM_011509129.1 | 3 | 236 | XP_011507431.1 | |
XM_011509130.1 | 3 | 236 | XP_011507432.1 | |
XM_011509131.1 | 3 | 236 | XP_011507433.1 | |
XM_011509132.2 | 3 | 236 | XP_011507434.1 | |
XM_011509133.2 | XP_011507435.1 | |||
XM_011509134.2 | XP_011507436.1 | |||
XM_011509135.2 | XP_011507437.1 | |||
XM_011509136.1 | 3 | 236 | XP_011507438.1 | |
XM_017000227.1 | XP_016855716.1 | |||
AF329488.1 | 3 | 175 | AAL23898.1 | |
AF459634.1 | 3 | 218 | AAL60250.1 | |
AK096690.1 | 3 | 204 | BAC04842.1 | |
AK315748.1 | 3 | 193 | ||
AL833970.1 | 1 | 190 | CAD38815.1 | |
AY043464.1 | 3 | 220 | AAK91777.1 | |
BC033690.1 | 3 | 216 | AAH33690.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |