Assay Details
Target Gene Details
Entrez Gene ID: | 1316 |
Gene Name: | Kruppel like factor 6 |
Gene Aliases: |
BCD1, CBA1, COPEB, CPBP, GBF, PAC1, ST12, ZF9 |
Location: |
Chr.10:3775996-3785281 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KLF6 | NM_001160124.1 | 2 | 590 | NP_001153596.1 |
NM_001160125.1 | 2 | 590 | NP_001153597.1 | |
NM_001300.5 | 2 | 590 | NP_001291.3 | |
NR_027653.1 | 2 | 590 | ||
AB017493.1 | 2 | 440 | BAA33050.1 | |
AF001461.1 | 2 | 353 | AAC39929.1 | |
AK293259.1 | 2 | 524 | ||
AK316596.1 | 2 | 518 | ||
BC000311.2 | 2 | 509 | AAH00311.1 | |
BM544849.1 | 2 | 591 | ||
BM695496.1 | 1 | 166 | ||
BP357332.1 | ||||
BT006778.1 | 2 | 323 | AAP35424.1 | |
CR975091.1 | 2 | 507 | ||
CR994352.1 | 2 | 507 | ||
FJ619623.1 | 2 | 330 | ||
U44975.1 | 2 | 344 | AAC23699.1 | |
U51869.1 | 2 | 675 | AAB94792.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv661n100 | Chr.10:3749938 - 3788015 on Build GRCh38 | Gain | KLF6 LOC105376360 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |