Assay Details
Target Gene Details
Entrez Gene ID: | 84172 |
Gene Name: | RNA polymerase I subunit B |
Gene Aliases: |
RPA135, RPA2, Rpo1-2 |
Location: |
Chr.2:112541915-112577153 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
POLR1B | NM_001137604.2 | 1 | 451 | NP_001131076.1 |
NM_001282772.1 | NP_001269701.1 | |||
NM_001282774.1 | 1 | 451 | NP_001269703.1 | |
NM_001282776.1 | 1 | 451 | NP_001269705.1 | |
NM_001282777.1 | 1 | 451 | NP_001269706.1 | |
NM_001282779.1 | 1 | 451 | NP_001269708.1 | |
NM_019014.5 | 1 | 451 | NP_061887.2 | |
AK128044.1 | 1 | 451 | BAC87247.1 | |
AK302814.1 | ||||
BC110833.1 | 1 | 140 | AAI10834.1 | |
BX647360.1 | 1 | 177 |
More Information
Additional Information:
For this assay, SNP(s) [rs138896228] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
Intragenic Exonic non-DGV Variation |