Assay Details
Target Gene Details
Entrez Gene ID: | 79143 |
Gene Name: | membrane bound O-acyltransferase domain containing 7 |
Gene Aliases: |
BB1, LENG4, LPIAT, LRC4, MBOA7, OACT7, hMBOA-7 |
Location: |
Chr.19:54173412-54190050 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MBOAT7 | NM_001146056.2 | 3 | 785 | NP_001139528.1 |
NM_001146082.2 | 5 | 991 | NP_001139554.1 | |
NM_001146083.2 | 4 | 864 | NP_001139555.1 | |
NM_024298.4 | 5 | 991 | NP_077274.3 | |
XM_011527299.2 | 5 | 1195 | XP_011525601.1 | |
XM_011527300.2 | 5 | 586 | XP_011525602.1 | |
XM_017027296.1 | 5 | 1159 | XP_016882785.1 | |
AK055908.1 | 5 | 924 | BAB71043.1 | |
AK122789.1 | 6 | 709 | ||
AK293199.1 | 3 | 395 | ||
AK298689.1 | 3 | 471 | ||
AK311493.1 | 5 | 628 | ||
BC002512.2 | 4 | 441 | AAH02512.2 | |
BC003164.1 | 5 | 522 | AAH03164.2 | |
BC006309.2 | 5 | 781 | AAH06309.1 | |
BC015857.1 | 4 | 428 | ||
EU016381.1 | 4 | 443 | ||
S82470.1 | 3 | 300 | AAB37433.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1066425 | Chr.19:54182441 - 54237431 on Build GRCh38 | Gain | LILRB3 LILRA6 MBOAT7 TSEN34 RPS9 |
nsv9751 | Chr.19:54158885 - 54193528 on Build GRCh38 | Gain | LENG1 TMC4 MBOAT7 TSEN34 |
nsv2546 | Chr.19:54182614 - 54270671 on Build GRCh38 | Deletion | LILRB3 LILRA6 LILRB5 MBOAT7 TSEN34 RPS9 |
nsv953613 | Chr.19:54097214 - 54198733 on Build GRCh38 | Deletion | TFPT LENG1 PRPF31 TMC4 NDUFA3 MBOAT7 TSEN34 CNOT3 OSCAR |
esv2718812 | Chr.19:53184636 - 54322450 on Build GRCh38 | Deletion | MIR521-1 MIR518A2 MIR935 LOC105372457 MIR520D MIR520H LILRB3 MIR520C TMC4 ZNF765 MIR516B1 LOC284379 DPRX MIR1283-1 MIR524 MIR526B MIR372 MIR517B MIR519A1 MIR526A1 TPM3P9 MIR517A MIR519C ZNF813 MIR371A MIR4752 PRPF31 VN1R2 NDUFA3 MIR373 MIR516B2 NLRP12 MIR515-1 MIR518E CACNG8 MIR525 MIR518F ZNF677 ZNF845 MIR523 LENG1 MIR371B MIR1283-2 MIR526A2 MIR518B MIR520B MIR518D MIR516A1 CACNG7 VSTM1 MIR516A2 LILRB5 MIR520F CNOT3 OSCAR ZNF525 MIR520G MIR519B BIRC8 MIR519E MIR512-1 MIR520A ZNF665 MBOAT7 ZNF761 TARM1 PRKCG MIR518A1 MIR515-2 MIR527 MIR520E MIR519D MYADM MIR519A2 LILRB2 TSEN34 MIR522 MIR521-2 MIR518C MIR512-2 CACNG6 TFPT MIR1323 ZNF331 LILRA6 VN1R4 MIR517C LILRA5 ZNF818P MIR498 FAM90A27P RPS9 |
nsv510774 | Chr.19:54158011 - 54269511 on Build GRCh38 | Deletion | LENG1 LILRB3 TMC4 LILRA6 LILRB5 MBOAT7 TSEN34 RPS9 |
esv2758771 | Chr.19:54163847 - 54954956 on Build GRCh38 | Gain+Loss | KIR2DL1 LENG8-AS1 LILRA4 LILRB3 TMC4 LENG8 LILRP2 LILRB5 NCR1 KIR3DX1 LILRA1 LILRB4 MBOAT7 LAIR2 LAIR1 NLRP7 LENG9 KIR2DL4 MIR4752 LILRA2 KIR3DL1 LILRB2 MIR8061 TSEN34 KIR3DL3 LOC102724300 LOC101928804 LILRB1 CDC42EP5 LILRA6 LILRA5 FCAR KIR2DS4 KIR2DL3 TTYH1 RPS9 KIR3DL2 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |