Assay Details
Target Gene Details
Entrez Gene ID: | 9940 |
Gene Name: | deleted in lung and esophageal cancer 1 |
Gene Aliases: |
CFAP81, DLC-1, DLC1, F56 |
Location: |
Chr.3:38038595-38122741 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DLEC1 | NM_001321153.1 | 7 | 1226 | NP_001308082.1 |
NM_007335.3 | 7 | 1226 | NP_031361.2 | |
NM_007337.3 | 7 | 1226 | NP_031363.2 | |
XM_006713438.3 | 7 | 1835 | XP_006713501.1 | |
XM_006713439.3 | 7 | 1834 | XP_006713502.1 | |
XM_006713440.3 | 7 | 1835 | XP_006713503.1 | |
XM_011534317.2 | 7 | 1834 | XP_011532619.1 | |
XM_011534318.2 | 7 | 1835 | XP_011532620.1 | |
AB020522.1 | 7 | 1226 | BAA77247.1 | |
AB425198.1 | 7 | 1198 | ||
AY789462.1 | 3 | 143 | AAX18633.1 | |
AY789464.1 | 3 | 167 | AAX18635.1 | |
BC129824.1 | 7 | 1226 | ||
BC129825.1 | 7 | 1226 | ||
BC171799.1 | 7 | 1226 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3383943 | Chr.3:38084209 - 38084703 on Build GRCh38 | Duplication | DLEC1 |
nsv508911 | Chr.3:38019295 - 38091072 on Build GRCh38 | Insertion | PLCD1 DLEC1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |