More Information
Set Membership: |
Non-coding |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein import into nucleus, translocation
osteoblast differentiation
maternal placenta development
positive regulation of protein phosphorylation
positive regulation of endothelial cell proliferation
glycogen biosynthetic process
regulation of glycogen biosynthetic process
glucose metabolic process
translation
regulation of translation
cellular protein modification process
protein phosphorylation
negative regulation of protein kinase activity
nitric oxide biosynthetic process
activation-induced cell death of T cells
inflammatory response
cellular response to DNA damage stimulus
response to oxidative stress
signal transduction
G-protein coupled receptor signaling pathway
germ cell development
aging
cell proliferation
insulin receptor signaling pathway
apoptotic mitochondrial changes
response to heat
negative regulation of autophagy
negative regulation of gene expression
negative regulation of plasma membrane long-chain fatty acid transport
positive regulation of fibroblast migration
positive regulation of sodium ion transport
positive regulation of glucose metabolic process
negative regulation of endopeptidase activity
regulation of neuron projection development
regulation of phosphatidylinositol 3-kinase signaling
glucose transport
phosphorylation
protein ubiquitination
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
spinal cord development
cell projection organization
cell differentiation
protein catabolic process
platelet activation
hyaluronan metabolic process
positive regulation of cell growth
regulation of cell migration
endocrine pancreas development
T cell costimulation
regulation of myelination
positive regulation of cyclin-dependent protein serine/threonine kinase activity involved in G1/S transition of mitotic cell cycle
lipopolysaccharide-mediated signaling pathway
negative regulation of fatty acid beta-oxidation
positive regulation of endodeoxyribonuclease activity
response to food
positive regulation of cellular protein metabolic process
peripheral nervous system myelin maintenance
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
cellular response to insulin stimulus
positive regulation of peptidyl-serine phosphorylation
response to fluid shear stress
intracellular signal transduction
cellular response to vascular endothelial growth factor stimulus
ERBB2 signaling pathway
glucose homeostasis
positive regulation of apoptotic process
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
regulation of mRNA stability
protein kinase B signaling
positive regulation of blood vessel endothelial cell migration
positive regulation of nitric oxide biosynthetic process
positive regulation of fat cell differentiation
positive regulation of glycogen biosynthetic process
positive regulation of epidermal growth factor receptor signaling pathway
negative regulation of cell size
negative regulation of proteolysis
positive regulation of vasoconstriction
positive regulation of transcription from RNA polymerase II promoter
positive regulation of glucose import
negative regulation of JNK cascade
protein autophosphorylation
positive regulation of lipid biosynthetic process
insulin-like growth factor receptor signaling pathway
phosphatidylinositol-mediated signaling
regulation of nitric-oxide synthase activity
positive regulation of nitric-oxide synthase activity
positive regulation of sequence-specific DNA binding transcription factor activity
striated muscle cell differentiation
response to growth hormone
mammary gland epithelial cell differentiation
glycogen cell differentiation involved in embryonic placenta development
labyrinthine layer blood vessel development
response to UV-A
cellular response to mechanical stimulus
cellular response to epidermal growth factor stimulus
cellular response to prostaglandin E stimulus
cellular response to organic cyclic compound
cellular response to hypoxia
establishment of protein localization to mitochondrion
maintenance of protein location in mitochondrion
positive regulation of establishment of protein localization to plasma membrane
negative regulation of release of cytochrome c from mitochondria
cellular response to granulocyte macrophage colony-stimulating factor stimulus
execution phase of apoptosis
chemical synaptic transmission, postsynaptic
negative regulation of protein kinase activity by protein phosphorylation
positive regulation of protein localization to nucleus
negative regulation of neuron death
regulation of signal transduction by p53 class mediator
regulation of cell cycle checkpoint
negative regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
cellular response to nerve growth factor stimulus
response to insulin-like growth factor stimulus
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
protein kinase activity
protein serine/threonine kinase activity
protein serine/threonine/tyrosine kinase activity
protein kinase C binding
protein binding
ATP binding
phosphatidylinositol-3,4,5-trisphosphate binding
kinase activity
enzyme binding
nitric-oxide synthase regulator activity
GTPase activating protein binding
identical protein binding
phosphatidylinositol-3,4-bisphosphate binding
protein phosphatase 2A binding
14-3-3 protein binding
protein import into nucleus, translocation
osteoblast differentiation
maternal placenta development
positive regulation of protein phosphorylation
positive regulation of endothelial cell proliferation
glycogen biosynthetic process
regulation of glycogen biosynthetic process
glucose metabolic process
translation
regulation of translation
cellular protein modification process
protein phosphorylation
negative regulation of protein kinase activity
nitric oxide biosynthetic process
activation-induced cell death of T cells
inflammatory response
cellular response to DNA damage stimulus
response to oxidative stress
signal transduction
G-protein coupled receptor signaling pathway
germ cell development
aging
cell proliferation
insulin receptor signaling pathway
apoptotic mitochondrial changes
response to heat
negative regulation of autophagy
negative regulation of gene expression
negative regulation of plasma membrane long-chain fatty acid transport
positive regulation of fibroblast migration
positive regulation of sodium ion transport
positive regulation of glucose metabolic process
negative regulation of endopeptidase activity
regulation of neuron projection development
regulation of phosphatidylinositol 3-kinase signaling
glucose transport
phosphorylation
protein ubiquitination
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
spinal cord development
cell projection organization
cell differentiation
protein catabolic process
platelet activation
hyaluronan metabolic process
positive regulation of cell growth
regulation of cell migration
endocrine pancreas development
T cell costimulation
regulation of myelination
positive regulation of cyclin-dependent protein serine/threonine kinase activity involved in G1/S transition of mitotic cell cycle
lipopolysaccharide-mediated signaling pathway
negative regulation of fatty acid beta-oxidation
positive regulation of endodeoxyribonuclease activity
response to food
positive regulation of cellular protein metabolic process
peripheral nervous system myelin maintenance
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
cellular response to insulin stimulus
positive regulation of peptidyl-serine phosphorylation
response to fluid shear stress
intracellular signal transduction
cellular response to vascular endothelial growth factor stimulus
ERBB2 signaling pathway
glucose homeostasis
positive regulation of apoptotic process
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
regulation of mRNA stability
protein kinase B signaling
positive regulation of blood vessel endothelial cell migration
positive regulation of nitric oxide biosynthetic process
positive regulation of fat cell differentiation
positive regulation of glycogen biosynthetic process
positive regulation of epidermal growth factor receptor signaling pathway
negative regulation of cell size
negative regulation of proteolysis
positive regulation of vasoconstriction
positive regulation of transcription from RNA polymerase II promoter
positive regulation of glucose import
negative regulation of JNK cascade
protein autophosphorylation
positive regulation of lipid biosynthetic process
insulin-like growth factor receptor signaling pathway
phosphatidylinositol-mediated signaling
regulation of nitric-oxide synthase activity
positive regulation of nitric-oxide synthase activity
positive regulation of sequence-specific DNA binding transcription factor activity
striated muscle cell differentiation
response to growth hormone
mammary gland epithelial cell differentiation
glycogen cell differentiation involved in embryonic placenta development
labyrinthine layer blood vessel development
response to UV-A
cellular response to mechanical stimulus
cellular response to epidermal growth factor stimulus
cellular response to prostaglandin E stimulus
cellular response to organic cyclic compound
cellular response to hypoxia
establishment of protein localization to mitochondrion
maintenance of protein location in mitochondrion
positive regulation of establishment of protein localization to plasma membrane
negative regulation of release of cytochrome c from mitochondria
cellular response to granulocyte macrophage colony-stimulating factor stimulus
execution phase of apoptosis
chemical synaptic transmission, postsynaptic
negative regulation of protein kinase activity by protein phosphorylation
positive regulation of protein localization to nucleus
negative regulation of neuron death
regulation of signal transduction by p53 class mediator
regulation of cell cycle checkpoint
negative regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
cellular response to nerve growth factor stimulus
response to insulin-like growth factor stimulus
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
protein kinase activity
protein serine/threonine kinase activity
protein serine/threonine/tyrosine kinase activity
protein kinase C binding
protein binding
ATP binding
phosphatidylinositol-3,4,5-trisphosphate binding
kinase activity
enzyme binding
nitric-oxide synthase regulator activity
GTPase activating protein binding
identical protein binding
phosphatidylinositol-3,4-bisphosphate binding
protein phosphatase 2A binding
14-3-3 protein binding