Product Details
- Assay Reference Genome
Location
Chr.3:26331301 on build GRCm38
- Cytoband
- 3A3
- Species
- Mus musculus
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 192167
- Gene Symbol
- Nlgn1
- Gene Name
- neuroligin 1
- Gene Aliases
- 6330415N05Rik, BB179718, NL1, Nlg1, mKIAA1070
- Location
- Chr.3:25431841-26332056 on build GRCm38
- Assay Gene Location
- Within Exon 1
View Full Product Details