Product Details

Assay Reference Genome
Location

Chr.1:123333632 on build GRCm38
Cytoband
1E2.3
Species
Mus musculus
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
269109
Gene Symbol
Dpp10
Gene Name
dipeptidylpeptidase 10
Gene Aliases
6430601K09Rik, DPP X, Dprp3
Location
Chr.1:123332138-124845960 on build GRCm38
Assay Gene Location
Within Exon 26
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
Dpp10 NM_199021.3 26 3136 NP_950186.3
AK032579.1 24 2782
BC029696.1 11 1618 AAH29696.1
BC063074.1 16 2058 AAH63074.1
BC067026.1 26 3129 AAH67026.1

View Full Product Details