Product Details
- Assay Reference Genome
Location
Chr.19:22897651 on build GRCm38
- Cytoband
- 19B
- Species
- Mus musculus
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 226025
- Gene Symbol
- Trpm3
- Gene Name
- transient receptor potential cation channel, subfamily M, member 3
- Gene Aliases
- 6330504P12Rik, 9330180E14, AU018608, B930001P07Rik, LTRPC3, MLSN2
- Location
- Chr.19:22138181-22989897 on build GRCm38
- Assay Gene Location
- Overlaps Intron 17 - Exon 18
View Full Product Details