Product Details
- Assay Reference Genome
Location
Chr.16:72972553 on build GRCm38
- Cytoband
- 16C3.1
- Species
- Mus musculus
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 19876
- Gene Symbol
- Robo1
- Gene Name
- roundabout homolog 1 (Drosophila)
- Gene Aliases
- AW494633, AW742721, DUTT1, Gm310
- Location
- Chr.16:72027553-73047491 on build GRCm38
- Assay Gene Location
- Within Exon 12
View Full Product Details