Product Details
- Assay Reference Genome
Location
Chr.15:22934884 on build GRCm38
- Cytoband
- 15A2
- Species
- Mus musculus
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 320865
- Gene Symbol
- Cdh18
- Gene Name
- cadherin 18
- Gene Aliases
- B230220E17Rik, Cdh14l
- Location
- Chr.15:22548899-23474432 on build GRCm38
- Assay Gene Location
Gene Symbol |
Transcript Accession |
Exon Location |
Assay Location |
Protein ID |
Cdh18 |
AK045672.1 |
|
|
BAC32451.1 |
View Full Product Details