Product Details
- Assay Reference Genome
Location
Chr.15:23382867 on build GRCm38
- Cytoband
- 15A2
- Species
- Mus musculus
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 320865
- Gene Symbol
- Cdh18
- Gene Name
- cadherin 18
- Gene Aliases
- B230220E17Rik, Cdh14l
- Location
- Chr.15:22548899-23474432 on build GRCm38
- Assay Gene Location
- Within Exon 6
View Full Product Details